Learn More
Ergodyne™ Chill-Its™ 6705CT Evaporative Hook and Loop Bandana with Cooling Towel
Shop All Ergodyne ProductsDescription
- Advanced PVA material activates easily and quickly when run under water for one minute
- Reusable
- Super-evaporative cooling process helps keep you cool
- Breakaway hook and loop closure
- Available in a variety of colors and patterns like red western, blue western, camo, stars and striped, black, lime, blue, skulls, and flames
Specifications
Specifications
| Material | PVA |
| Product Type | Bandana |
| Color | Red, White |
| Closure Type | Hook and Loop |
| For Use With (Application) | Construction, Maintenance, Construction, Landscaping, Material Handling |
| Includes | Cooling Towel |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.