Learn More
Description
- Activate by soaking in water for 2-5 minutes
- Reusable
- Tie closure
- Available in a variety of colors and patterns like stars and stripes, camouflage, red western, navy western, blue, lime, flames, and skulls
Specifications
Specifications
| Product Type | Cooling Hat |
| Color | Red, White |
| Closure Type | Tie |
| For Use With (Application) | Construction, Maintenance, Landscaping, Material Handling |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.