Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ Chill-Its™ 6710 Evaporative Cooling Triangle Hat

Catalog No. 19819499 Shop All Ergodyne Products
Change view
Click to view available options
Color:
Black
Blue
Blue, White
Camouflage
Gray, Black, White
Lime
Red, Orange, Black
Red, White
Red, White, Blue
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-819-499 Black
19-819-487 Red, White, Blue
19-819-488 Camouflage
19-819-489 Red, White
19-819-490 Blue, White
19-819-495 Lime
19-819-491 Blue
19-819-493 Red, Orange, Black
19-819-507 Gray, Black, White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 19-819-499 Supplier Ergodyne™ Supplier No. 12335

Evaporative Cooling Triangle Hat features activated polymers that provide cooling power in a variety of applications.

  • Activate by soaking in water for 2-5 minutes
  • Reusable
  • Tie closure
  • Available in a variety of colors and patterns like stars and stripes, camouflage, red western, navy western, blue, lime, flames, and skulls

Specifications

Product Type Cooling Hat
Color Black
For Use With (Application) Construction, Maintenance, Landscaping, Material Handling
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.