Learn More
Ergodyne™ Chill-Its™ 6710CT Evaporative Cooling Triangle Hat with Cooling Towel
Shop All Ergodyne ProductsDescription
- Advanced PVA material activates easily and quickly once run under water for one minute
- Reusable
- Super-evaporative cooling process helps keep you cool
- Lightweight and low profile
- Tie closure
- Available in a variety of patterns and colors like blue, lime, red western, blue western, stars and stripes, flames, camouflage, and skulls
Specifications
Specifications
| Material | PVA |
| Product Type | Cooling Hat |
| Color | Red, White |
| For Use With (Application) | Construction, Maintenance, Landscaping, Material Handling |
| Includes | Cooling Towel |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.