Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ Chill-Its™ 6710CT Evaporative Cooling Triangle Hat with Cooling Towel

Catalog No. 19819571 Shop All Ergodyne Products
Change view
Click to view available options
Color:
Black
Blue
Blue, White
Camouflage
Gray, Black, White
Lime
Red, Orange, Black
Red, White
Red, White, Blue
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-819-571 Blue
19-819-565 Red, White, Blue
19-819-566 Camouflage
19-819-567 Red, White
19-819-570 Lime
19-819-572 Red, Orange, Black
19-819-573 Gray, Black, White
19-819-568 Blue, White
19-819-569 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 19-819-571 Supplier Ergodyne™ Supplier No. 12587
Add to Cart
Add to Cart

Evaporative Cooling Triangle Hat with Cooling Towel is constructed of advanced PVA material designed to keep you cool without the bulk.

  • Advanced PVA material activates easily and quickly once run under water for one minute
  • Reusable
  • Super-evaporative cooling process helps keep you cool
  • Lightweight and low profile
  • Tie closure
  • Available in a variety of patterns and colors like blue, lime, red western, blue western, stars and stripes, flames, camouflage, and skulls

Specifications

Material PVA
Product Type Cooling Hat
Color Blue
Closure Type Tie
For Use With (Application) Construction, Maintenance, Landscaping, Material Handling
Includes Cooling Towel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.