Learn More
Thermo Scientific™ Chiller Plumbing Kits and Accessories
Description
Thermo Scientific™ Chiller Plumbing Kits and accessories include flexible tubing, tubing insulation, and hose clamps.
Features:
- Include flexible tubing, tubing insulation, and hose clamps
Specifications
Specifications
| For Use With (Equipment) | ThermoFlex 900 to 10000 |
| Type | Plumbing Package |
| Includes | 25 ft. of 5/8 in. ID high temp hose, insulation, hose barbs and clamps |
| Material | Material for High Temperature |
| Size | 1/2 in. NPT, 25 ft. |