Learn More
Thermo Scientific™ Chiller Plumbing Kits and Accessories
Description
Thermo Scientific™ Chiller Plumbing Kits and accessories include flexible tubing, tubing insulation, and hose clamps.
Features:
- Include flexible tubing, tubing insulation, and hose clamps
Specifications
Specifications
| For Use With (Equipment) | ThermoTemp, Polar, VersaCool |
| Type | Tubing |
| Includes | Insulated flexible SS tubing with M16x1 threaded fittings 1.0 meter long |
| Material | Stainless-steel |
| Size | 6 mm ID per meter, 1.0 meter long |