Learn More
Thermo Scientific™ Chiller Plumbing Kits and Accessories
Description
Thermo Scientific™ Chiller Plumbing Kits and accessories include flexible tubing, tubing insulation, and hose clamps.
Features:
- Include flexible tubing, tubing insulation, and hose clamps
Specifications
Specifications
| For Use With (Equipment) | ThermoFlex 3500 to 5000 |
| Type | Plumbing Package |
| Includes | 32.8 ft. (10 meters) of 5/8 in. ID Polybraid hose (use with 3/4 in. hose barbs) , insulation and clamps |
| Material | Polybraid |
| Size | 3/4 in. NPT, 32.8 ft. |