Learn More
Active Motif ChIP-IT Control Kit – Human
SDP
Supplier:  Active Motif 53010
Encompass_Preferred
ChIP-IT Kits provide all of the critical components needed for successful ChIP in a single kit. ChIP-IT comes complete with positive & negative control antibodies and PCR primers, DNA purification columns, optimized buffers and a comprehensive manual, all of which have been proven to work in ChIP. For your convenience, kits are available in a variety of formats, with your choice of sonication or our exclusive enzymatic shearing, and either with or without positive & negative controls. This enables you to use the shearing method that works best for you, and to either receive human-specific controls with the kit or to purchase ChIP-IT Control Kits separately to help analyze your results and verify that your antibodies work in the species you choose.
Catalog No. NC9522386
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.