Learn More
Active Motif ChIP-IT FFPE II
Supplier: Active Motif 53047
Formalin-fixed paraffin-embedded (FFPE) clinical samples are a valuable resource for retrospective research. However, working with FFPE samples presents a challenge for epigenetics techniques such as ChIP. Active Motif has leveraged its epigenetics expertise to develop commercially available kits to enable ChIP from FFPE tissue for use in Next Generation Sequencing (ChIP-Seq). The FFPE ChIP assay comes in two modules, the Chromatin Preparation Kit (for which there are two options (Cat. Nos. 53030 & 53031), depending on sample quality) and the ChIP-IT FFPE II Kit (Cat. No. 53047) to perform the ChIP-based enrichment. This enables you to work with different types and amounts of starting material.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.