Learn More
Eurofins Discoverx CHIR 99021 (CT99021), 2 mg. (92-1116)
Supplier:  Eurofins Discoverx 921116
Encompass_Preferred
CHIR-99021 HCl is a synthetic small molecule and ATP-competitive inhibitor of GSK3 kinase, with IC50 of 10 and 6.7 nM for GSK3α and GSK3β, respectively. Has >500-fold selectivity for GSK3 over closely related CDC2 and ERK2 kinases. Functions as a WNT activator. Provided as 2 mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9650
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.