Learn More
Chiral Technologies, Inc. ChiralPak IC-3/SFC Chiral Column, Universal Solvent Compatibility, 3.0 mm Diameter
Supplier:  Chiral Technologies, Inc. 83S84
Encompass
The CHIRALPAK IC-3/SFC 3mm x 150mm column from Daicel Chiral Technologies is a highly efficient analytical chiral column designed for Supercritical Fluid Chromatography (SFC) and compatible with UHPLC. This column offers exceptional selectivity for enantiomer resolution and robust solvent compatibility, enabling diverse mobile phase selections. With its immobilized polysaccharide stationary phase and high pressure stability, it ensures reliable performance across various applications, setting it apart from other chiral columns on the market.
Catalog No. NC2601517
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.