Learn More
Chiral Technologies, Inc. CHIRALPAK IM 4.6X250MM 5µM, High-Performance HPLC Column, Universal Solvent Compatibility
Supplier:  Chiral Technologies, Inc. 92325
Encompass
The CHIRALPAK IM 4.6x250mm 5µm is an advanced analytical column designed for high-performance liquid chromatography (HPLC) and supercritical fluid chromatography (SFC). It offers enhanced robustness with its immobilized polysaccharide chiral stationary phase, enabling exceptional resolution for enantiomeric separations. Its universal solvent compatibility allows for innovative selectivity profiles, making it a distinct choice for challenging separations compared to conventional chiral columns.
Catalog No. NC2326273
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.