Learn More
Invitrogen™ Chk1 Polyclonal Antibody

Description
Positive Samples: SW620, HT-29, Mouse thymus, Mouse testis, Rat testis; Cellular Location: Cytoplasm, Nucleus, centrosome, cytoskeleton, microtubule organizing center Immunogen sequence: DFGLATVFRY NNRERLLNKM CGTLPYVAPE LLKRREFHAE PVDVWSCGIV LTAMLAGELP WDQPSDSCQE YSDWKEKKTY LNPWKKIDSA PLALLHKILV ENPSARITIP DIKKDRWYNK PLKKGAKRPR VTSGGVSESP SGFSKHIQSN LDFSPVNSAS.
Specifications
Specifications
| Antigen | Chk1 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.35 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 |
| Gene | Chek1 |
| Gene Accession No. | O14757, O35280, Q91ZN7 |
| Gene Alias | C85740; Cell cycle checkpoint kinase; checkpoint kinase 1; checkpoint kinase 1 homolog; checkpoint kinase-1; Checkpoint, S. pombe, homolog of, 1; CHEK1; CHK1; CHK1 checkpoint homolog; Chk1-S; homolog of Chk1-S; HRD1 Protein; rad27; rad27 homolog; serine/threonine protein kinase CHK1; serine/threonine-protein kinase Chk1; Synoviolin; YBR1742; YBR274W |
| Gene Symbols | Chek1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.