Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHKB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112385
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112385 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112385 Supplier Thermo Scientific Supplier No. PA5112385
Only null left
Add to Cart
Add to Cart

CHKB Polyclonal Antibody

CHKB Polyclonal Antibody for Western Blot, IHC (P), ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen CHKB
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene CHKB
Gene Accession No. O54783, O55229, Q9Y259
Gene Alias Chetk; CHKB; Chkl; choline kinase beta; Choline kinase-like protein; choline/ethanolamine kinase; choline/ethanolamine kinase beta; choline/ethanolamine kinase-b; choline/ethanolaminekinase; CK; Ck/Ek; Ck/Ek-beta; CKB; CKEKB; EK; EKB; ethanolamine kinase; Ethanolamine kinase beta; MDCMC
Gene Symbols CHKB
Host Species Rabbit
Immunogen Recombinant Human Choline/ethanolamine kinase protein (2-99AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1120, 12651, 29367
Target Species Human, Mouse, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.