Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ CHL-1/L1CAM-2 Antibody Pair [HRP]

Mouse Monoclonal Antibody Pair
$1195.00
Specifications
Assay Range | 31.25 pg/mL |
---|---|
Assay Sensitivity | 31.25-2000 pg/mL |
Assay | Sandwich ELISA |
Conjugate | HRP |
Product Type | Antibody Pairs |
Description
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human CHL-1/L1CAM-2.Specifications
31.25 pg/mL | |
Sandwich ELISA | |
Antibody Pairs | |
Human | |
CALLClose homolog of L1, cell adhesion molecule with homology to L1CAM (close homolog of L1), cell adhesion molecule with homology to L1CAM (close homologue of L1), FLJ44930, L1CAM2L1 cell adhesion molecule 2, MGC132578, neural cell adhesion molecule L1-like protein | |
CHL1 | |
Storage is content dependent. |
31.25-2000 pg/mL | |
HRP | |
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human CHL-1/L1CAM-2. | |
Sandwich ELISA | |
10752 | |
Mouse | |
RUO | |
CHL-1/L1CAM-2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title