Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHL1 Monoclonal Antibody (05), Invitrogen™

Mouse Monoclonal Antibody
$300.00 - $516.00
Specifications
| Antigen | CHL1 |
|---|---|
| Clone | 05 |
| Concentration | 1 mg/mL |
| Applications | ELISA |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
PIMA543402
|
Invitrogen™
MA543402 |
50 μL |
Each for $300.00
|
|
|||||
PIMA542976
![]() |
Invitrogen™
MA542976 |
100 μL |
Each for $516.00
|
|
|||||
Description
Specificity: Human CHL1/LICAM2/CALL
The protein encoded by this gene is a member of the L1 gene family of neural cell adhesion molecules. It is a neural recognition molecule that may be involved in signal transduction pathways. The deletion of one copy of this gene may be responsible for mental defects in patients with 3p- syndrome. Several alternatively spliced transcript variants of this gene have been described, but their full length nature is not known.Specifications
| CHL1 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative | |
| A530023M13Rik; AI465420; CALL; cell adhesion molecule L1 like; cell adhesion molecule L1-like; cell adhesion molecule with homology to L1CAM; cell adhesion molecule with homology to L1CAM (close homolog of L1); cell adhesion molecule with homology to L1CAM (close homologue of L1); Chl1; Chl1-like protein; close homolog of L1; L1 cell adhesion molecule 2; L1CAM2; LICAM2; neural cell adhesion molecule L1-like protein; Processed neural cell adhesion molecule L1-like protein | |
| CHL1 | |
| IgG1 | |
| Protein A | |
| Antibody |
| 05 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| O00533 | |
| 10752 | |
| Recombinant Human CHL1/LICAM2/CALL Protein. | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| CHL1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title