Learn More
Invitrogen™ Chlamydia trachomatis EBs Polyclonal Antibody

Description
PA1-7208 detects the Chlamydia trachomatis. This antibody cross reacts with Chlamydia psittacii and Chlamydia pneumoniae (TWAR). PA1-7208 has been successfully used in immunofluorescence procedures. The PA1-7208 immunogen is L2 and other serovar groups. Recommended storage of up to 6 months at 2-8°C. For long term storage, aliquot and freeze. avoiding freeze/thaw cycles.
Specifications
Specifications
| Antigen | Chlamydia trachomatis EBs |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 5.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.2 |
| Gene Alias | C. trachomatis; C. trachomatis EBS; Chlamydia EB; Chlamydia EBs |
| Host Species | Goat |
| Immunogen | L2 and other serovar groups. |
| Purification Method | IgG fraction |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.