Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloramine B Hydrate 80.0+%, TCI America™
SDP

Catalog No. C007525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C007525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C007525G Supplier TCI America Supplier No. C007525G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 127-52-6
Molecular Formula C6H5ClNNaO2S
Molecular Weight (g/mol) 213.61
MDL Number MFCD01459908
InChI Key FTDKYSGFGRDGGE-UHFFFAOYSA-M
Synonym Benzenesulfonechloramide Sodium Salt
PubChem CID 22141507
IUPAC Name sodium (chloroimino)(phenyl)-λ⁶-sulfanoylolate
SMILES [Na+].[O-]S(=O)(=NCl)C1=CC=CC=C1

Specifications

Color White-Yellow
Quantity 25 g
UN Number 1479
Formula Weight 213.61
Percent Purity ≥80.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Chloramine B Hydrate
RTECSNumber DA9300000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.