Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloramine-T Trihydrate, Reagent, ACS, 98-103%, Spectrum™ Chemical
SDP

Catalog No. 18601583 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
125 g
250 g
500 g
2.5 kg
Packaging:
Amber Glass Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-583 2.5 kg Poly Pail
18-601-581 125 g Amber Glass Bottle
18-601-585 250 g Amber Glass Bottle
18-601-582 500 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-601-583 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. CH1202.5KG
Add to Cart
Add to Cart

CH120, 7080-50-4, C7H7SO2NClNa·3H2O

Spectrum™ Chemical Chloramine-T Trihydrate, Reagent, ACS is also known as tosylchloramide. It is most commonly used as a disinfectant or as a biocide. It is a white powder and can be a source of electrophilic chlorine in organic synthesis. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

Molecular Formula C7H13ClNNaO5S
Molecular Weight (g/mol) 281.68
MDL Number MFCD00149066
InChI Key RNROOTWAJBXJBD-UHFFFAOYSA-M
IUPAC Name sodium (chloroimino)(4-methylphenyl)-λ⁶-sulfanoylolate trihydrate
SMILES O.O.O.[Na+].CC1=CC=C(C=C1)S([O-])(=O)=NCl

Specifications

CAS Max % 100%
Quantity 2.5 kg
UN Number UN3263
Percent Purity 98 to 103%
Grade ACS
Packaging Poly Pail
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.