Learn More
Abcam Chloramphenicol, Broad Spectrum Antibiotic, >99% Purity, 5 g
Description
- Molecular weight: 323.13 Da
- Purity: >99%
- Form: Solid
- Mechanism of action: Broad spectrum antibiotic agent that binds selectively to A2451 and A2452 residues in the 23S rRNA of the 50S ribosomal subunit, preventing peptide bond formation and protein chain elongation
- Biological effect: Inhibits protein biosynthesis and shows bacteriostatic effects against a wide variety of Gram-positive and Gram-negative bacteria in vivo
- Additional properties: Orally active
- Composition: 99.82% Chloramphenicol
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Content And Storage | Store at -20°C. |
| Shipping Condition | Blue Ice |
| Buffer | 99.82% chloramphenicol |
| Form | Solid |
| Target Molecule | 23S rRNA |
| Formulation | Constituents: 99.82% Chloramphenicol |
| Quantity | 5 g |
| For Use With (Application) | Protein biosynthesis inhibition |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.