Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloramphenicol Palmitate 97.0+%, TCI America™
SDP

Catalog No. C32581G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C32581G 1 g
C3258200MG 200 mg
C32585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C32581G Supplier TCI America Supplier No. C32581G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 530-43-8
Molecular Formula C27H42Cl2N2O6
Molecular Weight (g/mol) 561.541
MDL Number MFCD00083597
InChI Key PXKHGMGELZGJQE-ILBGXUMGSA-N
Synonym D-(-)-threo-2-Dichloroacetamido-1-(4-nitrophenyl)-1,3-propanediol Palmitate, 2,2-Dichloro-N-[(1R,2R)-1,3-dihydroxy-1-(4-nitrophenyl)-2-propyl]acetamide Palmitate
PubChem CID 443382
ChEBI CHEBI:3605
IUPAC Name [(2R,3R)-2-[(2,2-dichloroacetyl)amino]-3-hydroxy-3-(4-nitrophenyl)propyl] hexadecanoate
SMILES CCCCCCCCCCCCCCCC(=O)OCC(C(C1=CC=C(C=C1)[N+](=O)[O-])O)NC(=O)C(Cl)Cl

Specifications

Melting Point 94°C
Color White
Quantity 1 g
Formula Weight 561.54
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Chloramphenicol Palmitate
RTECSNumber RT4735000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.