Learn More
Research Products International Corp Chloramphenicol Sol 25 ml
SDP
Supplier:  Research Products International Corp ZA100225
Encompass_Preferred
Chloramphenicol inhibits bacterial protein synthesis by binding the 50S ribosomal subunit. Chloramphenicol is effective against both Gram (+) and (-) bacteria and some mycobacteria. Resistance to chloramphenicol is conferred by the cat gene which encodes an acetyltransferase that acetylates and inactivates the antibiotic. Chloramphenicol is commonly used for the amplification of vectors in Gram (-) bacteria.
Catalog No. 50-238-8442
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.