Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Chlorazol Black E, pure

Catalog No. AC404400250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC404400250 25 g Glass bottle
AC404401000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC404400250 Supplier Thermo Scientific Chemicals Supplier No. 404400250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
This item is not returnable. View return policy

CAS: 1937-37-7 | C34H25N9Na2O7S2 | 781.73 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1937-37-7
Molecular Formula C34H25N9Na2O7S2
Molecular Weight (g/mol) 781.73
MDL Number MFCD00066363
InChI Key OLVNAWLXZDRGPL-UHFFFAOYSA-L
PubChem CID 44135447
IUPAC Name disodium;4-amino-3-[[4-[4-[(2,4-diaminophenyl)diazenyl]phenyl]phenyl]diazenyl]-5-oxo-6-(phenylhydrazinylidene)naphthalene-2,7-disulfonate
SMILES C1=CC=C(C=C1)NN=C2C(=CC3=CC(=C(C(=C3C2=O)N)N=NC4=CC=C(C=C4)C5=CC=C(C=C5)N=NC6=C(C=C(C=C6)N)N)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Absorption Ratio 0.97 to 1
Infrared Spectrum Authentic
Lambda Max 597 to 602nm
Formula Weight 781.73
Grade Pure
Packaging Glass bottle
Color Black
Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material Chlorazol Black E, Pure
Hazard Statement GHS H Statement:
May cause cancer.
Suspected of damaging the unborn child.
Precautionary Statement GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Use personal protective equipment as required.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.