Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorhexidine Acetate, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18601588 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
50 kg
Packaging:
Fiber Drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-588 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-601-588 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. C406650KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

C4066, 56-95-1, C22H30Cl2N10·2C2H4O2

Spectrum™ Chemical Chlorhexidine Acetate, USP

Chemical Identifiers

CAS 56-95-1
Molecular Formula C26H38Cl2N10O4
Molecular Weight (g/mol) 625.56
MDL Number MFCD00150042
InChI Key WDRFFJWBUDTUCA-UHFFFAOYSA-N
IUPAC Name N'-(6-{[amino({amino[(4-chlorophenyl)amino]methylidene}amino)methylidene]amino}hexyl)-1-({amino[(4-chlorophenyl)amino]methylidene}amino)methanimidamide; bis(acetic acid)
SMILES CC(O)=O.CC(O)=O.NC(NC1=CC=C(Cl)C=C1)=NC(N)=NCCCCCCN=C(N)N=C(N)NC1=CC=C(Cl)C=C1

Specifications

CAS Max % 100%
Quantity 50 kg
Ignition Residue 0.15%
Loss on Drying 3.5%
Packaging Fiber Drum
UN Number UN2811
Percent Purity 98 to 102%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.