accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHOCH3OHONH2NHClNNH2NNNH2NNH2HNCl(E/Z)(E/Z)(E/Z)(E/Z)

CAS RN 56-95-1

IUPAC Name: N'-(6-{[amino({amino[(4-chlorophenyl)amino]methylidene}amino)methylidene]amino}hexyl)-1-({amino[(4-chlorophenyl)amino]methylidene}amino)methanimidamide; bis(acetic acid)
Synonyms: chlorhexidine diacetate
Molecular Weight (g/mol): 625.56
Molecular Formula: C26H38Cl2N10O4
InChi Key: WDRFFJWBUDTUCA-UHFFFAOYSA-N
SMILES: CC(O)=O.CC(O)=O.NC(NC1=CC=C(Cl)C=C1)=NC(N)=NCCCCCCN=C(N)N=C(N)NC1=CC=C(Cl)C=C1
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (2)
  • (1)
Grade 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)