Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorhexidine diacetate, >97%, MP Biomedicals™

Catalog No. ICN19136125 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19136125 25 g
ICN19136105 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19136125 Supplier MP Biomedicals, Inc Supplier No. 0219136125
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 56-95-1
Molecular Formula C26H38Cl2N10O4
Molecular Weight (g/mol) 625.56
MDL Number MFCD00150042
InChI Key WDRFFJWBUDTUCA-UHFFFAOYSA-N
Synonym chlorhexidine diacetate, chlorhexidine acetate, bactigras, hibitane diacetate, caswell no. 481e, chlorohexidine diacetate, 10,040 diacetate, chlorhexidine di acetate, chlorhexidine acetate van, unii-5908zuf22y
PubChem CID 43834499
IUPAC Name N'-(6-{[amino({amino[(4-chlorophenyl)amino]methylidene}amino)methylidene]amino}hexyl)-1-({amino[(4-chlorophenyl)amino]methylidene}amino)methanimidamide; bis(acetic acid)
SMILES CC(O)=O.CC(O)=O.NC(NC1=CC=C(Cl)C=C1)=NC(N)=NCCCCCCN=C(N)N=C(N)NC1=CC=C(Cl)C=C1

Specifications

Quantity 25 g
Percent Purity ≥97%
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.