Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorhexidine Diacetate, MP Biomedicals™

Catalog No. MP219136180 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219136180 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP219136180 Supplier MP Biomedicals, Inc Supplier No. 0219136180
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Effective against a wide range of bacteria, some fungi and some viruses. Chlorhexidine Diacetate, MP Biomedicals is a cationic broad-spectrum antimicrobial agent used as an antiseptic and disinfectant agent. It is effective against both Gram-positive and

Chlorhexidine is a cationic broad-spectrum antimicrobial agent belonging to the bis(biguanide) family. Its mechanism of action involves destabilization of the outer bacterial membrane. It is used primarily as a topical antiseptic/disinfectant in wound healing, at catheterization sites, in various dental applications and in surgical scrubs.

  • It is an agent to prevent gingivitis and was used to study the effects of antimicrobials on dental plaque
  • Commercial ophthalmic products have used this agent to replace thimerosal as a preservative; however, it can cause skin irritation
  • It has been used to study the inactivation of HIV and the immobilization of sperm
  • It was used to study effective skin antisepsis and disinfection of medical devices
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-95-1
Molecular Formula C26H38Cl2N10O4
Molecular Weight (g/mol) 625.56
MDL Number MFCD00150042
InChI Key WDRFFJWBUDTUCA-UHFFFAOYSA-N
Synonym chlorhexidine acetate hydrate 1:2:x, chlorhexidine diacetate salt monohydrate, chlorhexidine diacetate monohydrate, 1,1 inverted exclamation marka-hexamethylenebis 5-4-chlorophenyl biguanide diacetate salt, 4-chlorophenyl 6-4-chlorophenyl amino iminomethyl amino iminomethy l amino hexyl amino iminomethyl amino iminomethyl amine, acetic acid, acetic a cid, oxamethane
PubChem CID 43834499
IUPAC Name acetic acid;(1E)-2-[6-[[amino-[(E)-[amino-(4-chloroanilino)methylidene]amino]methylidene]amino]hexyl]-1-[amino-(4-chloroanilino)methylidene]guanidine;hydrate
SMILES CC(O)=O.CC(O)=O.NC(NC1=CC=C(Cl)C=C1)=NC(N)=NCCCCCCN=C(N)N=C(N)NC1=CC=C(Cl)C=C1

Specifications

Melting Point 151°C to 159°C
Quantity 100 g
Solubility Information Soluble in water (0.15g/10mL water with heat), in ethanol (50mg/mL - clear, faint yellow solution), alcohol, glycerol, propylene, glycol, polyethylene glycols.
Formula Weight 625.56g/mol
Percent Purity ≥97%
Concentration or Composition (by Analyte or Components) CriticalMicelle Concentration (CMC) 0.01% (w/v, 25°C) (Lit.)
Physical Form Powder
Recommended Storage Store at Room Temperature (15–30°C)
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.