Learn More
Sigma Aldrich Fine Chemicals Biosciences WilmadR screw-cap NMR tube1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z2719261EA
Encompass
Precision Wilmad NMR tubes are suitable for high field NMR spectroscopy. NMR tube has an open-top screw-thread cap with a PTfE/silicone septum that permits addition of reactants or samples via syringe through the cap. Also good for sample storage. Caps and septa are replaceable.Excellent for handling air-sensitive samples and in cell perfusion studies.5 mm diameter tubes have size 8-425 caps (replacement Z162566)10 mm diameter tubes have size 15-425 caps (replacement Z106518)
Catalog No. 50-290-0857
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.