Learn More
Chlorhexidine, G-Biosciences

Description
It has been used against an array of oral pathogens, with a high effectiveness toward gram-negative bacteria.
Specifications
Specifications
| Quantity | 5 g |
| Boiling Point | 641.4°C |
| Product Line | Gibco |
| Physical Form | Powder |
| Purity | >98% |
| Molecular Weight (g/mol) | 505.45 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.