Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorhexidine Gluconate Solution, BP, 19-21%, Spectrum™ Chemical
SDP

Catalog No. 18601591 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-591 4 L Amber Glass Bottle
18-601-589 100 mL Amber Glass Bottle
18-601-590 500 mL Amber Glass Bottle
18-601-592 20 L Amber Glass Bottle
18-601-593 200 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-601-591 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. CH1264LTGL
Add to Cart
Add to Cart

CH126, 18472-51-0, 55-56-1, C22H30Cl2N10·2C6H12O7

Spectrum™ Chemical Chlorhexidine Gluconate Solution, BP is mostly used as a topical disinfectant. Spectrum Chemical manufactured BP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 18472-51-0
Molecular Formula C34H54Cl2N10O14
Molecular Weight (g/mol) 897.76
MDL Number MFCD00083599
InChI Key YZIYKJHYYHPJIB-UUPCJSQJSA-N
IUPAC Name bis((2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoic acid); N'-(6-{[amino({amino[(4-chlorophenyl)amino]methylidene}amino)methylidene]amino}hexyl)-1-({amino[(4-chlorophenyl)amino]methylidene}amino)methanimidamide
SMILES OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C(O)=O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C(O)=O.NC(NC1=CC=C(Cl)C=C1)=NC(N)=NCCCCCCN=C(N)N=C(N)NC1=CC=C(Cl)C=C1

Specifications

CAS Max % 100%
Boiling Point 97°C
Packaging Amber Glass Bottle
Quantity 4 L
Percent Purity 19-21%
Grade BP
Physical Form Liquid
Chemical Name or Material Chlorhexidine Gluconate Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.