Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloride Color Reagent, Ricca Chemical
SDP

Catalog No. 194032 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
1940-32 1 L
194016 500 mL
1940-1 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 1940-32 Supplier Ricca Chemical Company Supplier No. 194032
Add to Cart
Add to Cart
This item is not returnable. View return policy

For Chloride Determination by the Automated Ferricyanide Method

Specifications

Chemical Name or Material Chloride Color Reagent
Color Orange-Yellow
CAS 7732-18-5, 67-56-1, 7782-61-8, 7697-37-2, 592-85-8, 9002-92-0
CAS Min % 84.0, 10.43, 2.87, 0.42, 0.06, 0.05
CAS Max % 88.0, 10.86, 2.98, 0.44, 0.06, 0.05
Grade Laboratory
Packaging Amber Glass Bottle
Quantity 1 L
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.