Learn More
Apexbio Technology LLC SR-717(Synonyms: SR717, SR 717 lithium salt, SR-717 lithium, STING agonist SR-717, non-nucleotide STING agonist SR-717), 10mg, CAS: 2375421-09-1.
SDP
Supplier:  Apexbio Technology LLC B846310
Encompass_Preferred
SR-717 (CAS 2375421-09-1) is a non-nucleoside stimulator of interferon genes (STING) agonist structurally related to cyclic GMP AMP (cGAMP) It activates innate immunity by inducing interferon-stimulated gene (ISG) expression through STING signaling In cellular assays using wild-type and cGAS-knockout THP1 cell lines SR-717 demonstrated EC50 values of 2 1 M and 2 2 M respectively Given its mechanism of STING activation SR-717 serves as a valuable tool for research in cancer biology and immune regulation
Catalog No. 50-000-60714
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.