Learn More
Apexbio Technology LLC Chlorin e6(Synonyms: Ce6, Chlorophyllin e6, Chlorophyll derivative e6, Chlorin-e6, Chlorin e-6), 250mg, CAS: 19660-77-6.
SDP
Supplier:  Apexbio Technology LLC B8314250
Encompass_Preferred
Chlorin e6 (Ce6 CAS 19660-77-6) is a second-generation photosensitizer utilized in photodynamic therapy (PDT) research with demonstrated anticancer activity when activated by laser irradiation Upon irradiation Ce6 generates reactive oxygen species mediating cytotoxic effects and cellular apoptosis In mouse models intravenous administration of Ce6 (2 5 10 mg/kg) combined with irradiation (50 200 J/cm ) resulted in complete elimination of implanted fibrosarcomas In clinical trials for bronchogenic superficial squamous cell carcinoma Ce6-based PDT (40 mg/m 100 J/cm ) produced complete response rates up to 82 9% underscoring its utility for cancer therapy studies
Catalog No. 50-000-60371
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.