Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PureAire Monitoring Systems Chlorine Dioxide 1 ppm Sensor Cell
SDP

Catalog No. 18222003
Change view
Click to view available options
Components Measured:
Chlorine Dioxide
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Components Measured
18-222-003 Chlorine Dioxide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-222-003 Supplier PureAire Monitoring Systems Supplier No. 90069
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PureAire Monitoring Systems Chlorine Dioxide (CLO2) 1 ppm Sensor Cell for use with PureAire Toxic Gas Monitors

  • Sensor mounted
  • 6-12 life expectancy, with maintenance
  • Calibration parts available, sensor can last up to 10 years
  • Other gases or remote sensors available
  • 2-year warranty

Specifications

Components Measured Chlorine Dioxide
For Use With (Equipment) PureAire Toxic Gas Monitors
Type Fixed Gas Detection Monitor
Measurement Range 0 to 1 ppm
Warranty 2 Year
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.