Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorine DPD Reagent 3 Recommended Storage: 2°C to 8°C (36°F to 46°F)., Reagents

Chlorine DPD Reagent 3, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Supplier:  Reagents Holdings Llc CC00440020F

Quantity 20 L

 View more versions of this product

Catalog No. 01-335-705


Only null left
Add to Cart

Description

Description

Specifications

Chemical Identifiers

7732-18-5 , 7664-93-9
18.02
water
H2O
XLYOFNOQVPJJNP-UHFFFAOYSA-N
O

Specifications

7732-18-5 , 7664-93-9
H2O
O
18.02
Cubitainer
20 L
Liquid
100°C
XLYOFNOQVPJJNP-UHFFFAOYSA-N
water
Reagent
1.0001
Non-sterile
Chlorine DPD Reagent 3
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.