Learn More
Cayman Chemical DeslorelInacetate 50mg
SDP
Supplier:  Cayman Chemical 3404150 MG
Encompass_Preferred
A synthetic non-steroidal estrogen prevents DMBA-induced mammary tumor formation in rats at 5 mg/animal s.c. prevents bone loss and uterine atrophy in ovariectomized rats
Catalog No. 50-294-6992
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.