Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloro[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene]silver 97.0+%, TCI America™
SDP

Catalog No. C23731G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C23731G 1 g
C2373200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C23731G Supplier TCI America Supplier No. C23731G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 873297-19-9
Molecular Formula C27H36AgClN2
Molecular Weight (g/mol) 531.917
InChI Key GNVDTUAJUVJOFQ-UHFFFAOYSA-M
PubChem CID 49788523
IUPAC Name 1,3-bis[2,6-di(propan-2-yl)phenyl]imidazole;chlorosilver
SMILES CC(C)C1=C(C(=CC=C1)C(C)C)N2C=CN([C]2)C3=C(C=CC=C3C(C)C)C(C)C.Cl[Ag]

Specifications

Color White
Quantity 1 g
Formula Weight 531.92
Percent Purity ≥97.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Chloro[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene]silver
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.