Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloro(ethyl)dimethylsilane 97.0+%, TCI America™
SDP

Catalog No. D013525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D013525G 25 g
D01355G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D013525G Supplier TCI America Supplier No. D013525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6917-76-6
Molecular Formula C4H11ClSi
Molecular Weight (g/mol) 122.667
MDL Number MFCD00009918
InChI Key AVDUEHWPPXIAEB-UHFFFAOYSA-N
Synonym ethyldimethylchlorosilane, chlorodimethylethylsilane, chloro ethyl dimethylsilane, dimethylethylchlorosilane, dimethylethylsilyl chloride, ethyldimethylsilyl chloride, chloro ethyl dimethylsilane #, silane, chlorodimethylethyl-,
PubChem CID 138863
IUPAC Name chloro-ethyl-dimethylsilane
SMILES CC[Si](C)(C)Cl

Specifications

Color Yellow
Boiling Point 88°C
Quantity 25 g
UN Number 2985
Formula Weight 122.67
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Chloro(ethyl)dimethylsilane [Dimethylethylsilylating Agent]
EINECSNumber (2)-2041
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.