Learn More
Medchemexpress LLC Chloro(triphenylphosphine)gold(I) | 14243-64-2 | MFCD00009588 | 99.9% | 494.7 g/mol | C18H15AuClP | 100 MG

Supplier: Medchemexpress LLC HYW020955100MG
Chloro(triphenylphosphine)gold(I) is a gold(I) coordination complex used as a reagent and precursor in organometallic synthesis, homogeneous catalysis, and research applications. It consists of a gold center coordinated by a triphenylphosphine ligand and a chloride ligand.
- Molecular formula C18H15AuClP and molecular weight 494.7 g/mol.
- Commonly supplied as a high-purity crystalline solid for research use.
- Used as a reagent and precursor in organometallic synthesis and homogeneous catalysis.
- Soluble in common organic solvents for solution-phase reactions.
- Classified as an irritant; follow appropriate safety and handling procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.