Learn More
Medchemexpress LLC Chloro(triphenylphosphine)gold(I) | 14243-64-2 | MFCD00009588 | 99.9% | 494.71 g·mol⁻¹ | C18H15AuClP | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW02095550MG
Encompass_Preferred
Chloro(triphenylphosphine)gold(I) is an organogold reagent (C18H15AuClP) commonly used in organic synthesis and coordination chemistry. It is typically a white to very pale-yellow crystalline powder and serves as a precursor or active species in gold(I)-mediated transformations and research applications.
  • High chemical purity suitable for research applications.
  • Useful precursor for preparing gold(I) complexes and catalysts.
  • Stable crystalline solid at ambient conditions.
  • Molecular weight 494.71 g·mol⁻¹; molecular formula C18H15AuClP.
  • Well characterized and widely used in synthetic and coordination chemistry.
Catalog No. 50-003-00886