Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroacetaldehyde dimethyl acetal, 98%, Thermo Scientific™

Catalog No. AA3661622
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3661622 100 g
AA3661614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3661622 Supplier Thermo Scientific Chemicals Supplier No. 3661622

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-97-2
Molecular Formula C4H9ClO2
Molecular Weight (g/mol) 124.56
MDL Number MFCD00000948
InChI Key CRZJPEIBPQWDGJ-UHFFFAOYSA-N
Synonym chloroacetaldehyde dimethyl acetal, dimethylchloroacetal, dimethyl chloroacetal, dimethyl chloracetal, ethane, 2-chloro-1,1-dimethoxy, 2-chloro-1,1-dimethoxy-ethane, chloroacetaldehyde, dimethyl acetal, 2-chloroacetaldehyde, dimethyl acetal, methyl chloroacetal, 1-chloro-2,2-dimethoxyethane
PubChem CID 60986
IUPAC Name 2-chloro-1,1-dimethoxyethane
SMILES COC(CCl)OC

Specifications

Melting Point <-73°C
Density 1.094g/mL
Boiling Point 128°C to 130°C
Flash Point 28° (82°F)
Refractive Index 1.4150
Quantity 100 g
UN Number UN1993
Beilstein 1733700
Formula Weight 124.57
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Chloroacetaldehyde dimethyl acetal
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.