Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroacetonitrile, 98+%

Catalog No. p-7020762
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1282522 100 g
AAA1282536 500 g
AAA12825A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1282522 Supplier Thermo Scientific Chemicals Supplier No. A1282522
Only null left
Add to Cart

CAS: 107-14-2 | C2H2ClN | 75.495 g/mol

Chloroacetonitrile is used in the electrochemical synthesis of cyanoacetic acid with carbon dioxide. It is involved in phase-transfer-catalyzed Darzen's condensation reaction with cyclohexanone. It is also used as an eluent additive in thermospray liquid chromatography/mass spectrometry. Further, it is used to prepare polysubstituted pyrido[1,2-a]benzimidazole by reacting with other reactant such as malononitrile, aromatic aldehyde and pyridine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chloroacetonitrile is used in the electrochemical synthesis of cyanoacetic acid with carbon dioxide. It is involved in phase-transfer-catalyzed Darzen′s condensation reaction with cyclohexanone. It is also used as an eluent additive in thermospray liquid chromatography/mass spectrometry. Further, it is used to prepare polysubstituted pyrido[1,2-a]benzimidazole by reacting with other reactant such as malononitrile, aromatic aldehyde and pyridine.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-14-2
Molecular Formula C2H2ClN
Molecular Weight (g/mol) 75.495
MDL Number MFCD00001885
InChI Key RENMDAKOXSCIGH-UHFFFAOYSA-N
Synonym chloroacetonitrile, chloracetonitrile, acetonitrile, chloro, chloromethyl cyanide, monochloroacetonitrile, chloroethanenitrile, monochloromethyl cyanide, usaf kf-5, alpha-chloroacetonitrile, acetonitrile, 2-chloro
PubChem CID 7856
ChEBI CHEBI:82416
IUPAC Name 2-chloroacetonitrile
SMILES C(C#N)Cl

Specifications

Density 1.195
Boiling Point 124°C to 126°C
Flash Point 56°C (132°F)
Assay Percent Range ≥98%
Refractive Index 1.423
Quantity 100 g
Beilstein 506028
Solubility Information Immiscible with water.
Formula Weight 75.5
Physical Form Liquid
Chemical Name or Material Chloroacetonitrile
Show More Show Less
Hazard Category H226-H301+H331-H310-H319
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P262-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P310-P311-P330-P363-P370+P378q-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: I
EINECSNumber 203-467-0
RTECSNumber AL8225000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.