Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorobenzene-d{5}, 99% (Isotopic), Thermo Scientific Chemicals

Catalog No. p-7059493
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4471206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4471206 Supplier Thermo Scientific Chemicals Supplier No. 4471206
Only null left
Add to Cart

Chlorobenzene-d5 is a halogenated D5 chlorobenzene for proteomics research. It is a precursor to deuterated dyes and rubbers. It is used as a high-boiling solvent in industrial applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3114-55-4
Molecular Formula C6H5Cl
Molecular Weight (g/mol) 117.587
MDL Number MFCD00000531
InChI Key MVPPADPHJFYWMZ-RALIUCGRSA-N
Synonym chlorobenzene-d5, chloro 2h5 benzene, 1-chloro 2 h? benzene, chlorobenzene d5, benzene-d5, chloro, 2h5 phenyl chloride, 1-chloro 2h5 benzene, benzene-d5-, chloro, chlorobenzene, perdeutero, chlorobenzene-d5, 99 atom % d
PubChem CID 575975
IUPAC Name 1-chloro-2,3,4,5,6-pentadeuteriobenzene
SMILES C1=CC=C(C=C1)Cl

Specifications

Physical Form Liquid
Quantity 5 g
Density 1.157 g/mL
Refractive Index 1.522
Flash Point 24°C (75°F)
Odor Aromatic
Boiling Point 130°C
UN Number UN1134
Beilstein 1866335
Solubility Information Not miscible with water.
Formula Weight 117.6
Percent Purity 99%
Chemical Name or Material Chlorobenzene-d5, Isotopic
Show More Show Less
Hazard Category H226-H332
Hazard Statement GHS H Statement
H226-H332
Flammable liquid and vapor.
Harmful if inhaled.
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: CHLOROBENZENE
EINECSNumber 221-482-0
TSCA No
Recommended Storage Ambient

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.