Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorobutanol, Anhydrous, NF, 98-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18601603 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-603 12 kg Poly Pail
18-601-601 125 g Amber Glass Bottle
18-601-602 500 g Amber Glass Bottle
18-601-604 50 kg Fiber Drum
18-601-600 25 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-601-603 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. CH12312KGBL
Add to Cart
Add to Cart
Request a Quote

CH123, 57-15-8, C4H7Cl3O

Spectrum™ Chemical Chlorobutanol, Anhydrous, NF is a very effective preservative in many pharmaceutical products for eyes, ears and nose, dental preparations. The anhydrous form is useful in formulations where the absence of water is desired. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 57-15-8
Molecular Formula C4H7Cl3O
Molecular Weight (g/mol) 177.45
InChI Key OSASVXMJTNOKOY-UHFFFAOYSA-N
IUPAC Name 1,1,1-trichloro-2-methylpropan-2-ol
SMILES CC(C)(O)C(Cl)(Cl)Cl

Specifications

CAS Max % 100%
Packaging Poly Pail
Quantity 12 kg
Percent Purity 98-100.5%
Grade NF
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.