Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

γ-Chlorobutyric Acid, MP Biomedicals

Catalog No. ICN201799 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN201799 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN201799 Supplier MP Biomedicals, Inc Supplier No. 0520179910

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 627-00-9
Molecular Formula C4H7ClO2
Molecular Weight (g/mol) 122.548
InChI Key IPLKGJHGWCVSOG-UHFFFAOYSA-N
Synonym 4-chlorobutyric acid, butanoic acid, 4-chloro, butyric acid, 4-chloro, gamma-chlorobutyric acid, 4-chlorobutanoicacid, gammachlorobutyric acid, 4-chloro-butyric acid, 4-chloro-n-butyric acid, .gamma.-chlorobutyric acid, 4-chloranylbutanoic acid
PubChem CID 12300
IUPAC Name 4-chlorobutanoic acid
SMILES C(CC(=O)O)CCl

Specifications

Quantity 10 g
Formula Weight 122.55
Chemical Name or Material γ-Chlorobutyric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.