Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorodifluoromethane Reference Material, Organic, Methanol Solvent, Ampule, 1 mL

Catalog No. USCFC1101
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
USCFC1101 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. USCFC1101 Supplier Agilent Technologies Supplier No. CFC1101
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 75-45-6 , 67-56-1
Molecular Formula CHClF2
Molecular Weight (g/mol) 86.47
MDL Number MFCD00000821
InChI Key VOPWNXZWBYDODV-UHFFFAOYSA-N
PubChem CID 6372
ChEBI CHEBI:82417
IUPAC Name chlorodifluoromethane
SMILES FC(F)Cl

Specifications

CAS Max % 99.9, 0.01
Analyte or Component Names Chlorodifluoromethane,methanol
Element Chlorodifluoromethane, Methanol
Solvent or Matrix Methanol
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 100μg/mL
Chemical Name or Material Chlorodifluoromethane Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.