Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorodimethylphenylsilane 96.0+%, TCI America™
SDP

Catalog No. D114725ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D114725ML 25 mL
D11475ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D114725ML Supplier TCI America Supplier No. D114725ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 768-33-2
Molecular Formula C8H11ClSi
Molecular Weight (g/mol) 170.71
MDL Number MFCD00000499
InChI Key KWYZNESIGBQHJK-UHFFFAOYSA-N
Synonym chlorodimethylphenylsilane, chlorodimethyl phenyl silane, phenyldimethylchlorosilane, silane, chlorodimethylphenyl, dimethylphenylchlorosilane, chloro dimethyl phenylsilane, phenyl dimethylchlorosilane, chloro-dimethyl-phenyl-silane, benzene, chlorodimethylsilyl, pubchem10814
PubChem CID 13029
IUPAC Name chlorodimethylphenylsilane
SMILES C[Si](C)(Cl)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 198°C
Quantity 25 mL
UN Number 2987
Formula Weight 170.71
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Chlorodimethylphenylsilane
EINECSNumber (3)-2641
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.