Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroethane Solution, 1 Analyte, 100μg/mL in Methanol, Ultra Scientific

Catalog No. USHC0601
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-HC-060-1 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-HC-060-1 Supplier Agilent Technologies Supplier No. HC0601
May include imposed supplier surcharges.
 Restricted Item
Add to Cart
Add to Cart

Chemical Identifiers

CAS 75-00-3 , 67-56-1
Molecular Formula C2H5Cl
Molecular Weight (g/mol) 64.512
InChI Key HRYZWHHZPQKTII-UHFFFAOYSA-N
PubChem CID 6337
ChEBI CHEBI:47554
IUPAC Name chloroethane
SMILES CCCl

Specifications

Physical Form Liquid
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 100μg/mL
Chemical Name or Material Chloroethane, 100μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.