Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroform, ≥99.5%, Solstice

Catalog No. 60048404 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-048-404 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-048-404 Supplier Solstice Supplier No. 256701L

missing translation for 'validMainframeMsg'

for IR spectroscopy, contains ∼0.5% ethanol as stabilizer,≥99.5%

  • Solstice has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-66-3
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
MDL Number MFCD00000826
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
PubChem CID 6212
ChEBI CHEBI:35255
IUPAC Name chloroform
SMILES ClC(Cl)Cl

Specifications

Melting Point -63°C
Color Colorless
Density 1.492g/mL at 25°C
Boiling Point 60°C to 62°C
Packaging Glass bottle
Refractive Index 1.446
Quantity 1 L
UN Number UN1888
Beilstein 1731042
Vapor Density 4.1 (vs air)
Vapor Pressure 160mmHg (20°C)
Formula Weight 119.38g/mol
Percent Purity ≥99.5%
Chemical Name or Material Chloroform, Contains≈0.5% ethanol as stabilizer, for IR spectroscopy
Show More Show Less
Hazard Statement P261-P281-P305 + P351 + P338-P311
Precautionary Statement H302-H315-H319-H331-H336-H351-H361d-H372
DOTInformation 6.1/PGIII
EINECSNumber 200-663-8
RTECSNumber FS9100000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.